Learn More
20 20 Genesystems Inc BIOCHECK POWDER SCREEN 25EA/CS
Supplier:  20 20 Genesystems Inc GB100125EA/CS
Encompass_Preferred
The BioCheck Powder Screening Test Kit is used by first responders as the initial step to investigate suspicious powders which may contain bioterrorism agents. The kit provides a cost effective, reliable, screening tool to determine whether or not a powder includes biological material. This helps to rule out many of the benign substances that cause powder incidents (e.g. talcum powder, dry wall dust, powdered sugar, etc.) that citizens have frequently feared to be possible biological weapons. As a result, this test allows first responders to screen for the possible presence of multiple bioterrorism agents such as anthrax and ricin toxin, while ruling out most ordinary substances. The product is now a primary screening test used by more than 500 fire departments and other first responders including a dozen federal agencies.
Catalog No. NC9976765