Learn More
Biochain Institute Inc Phosphate Assay Kit, 500 Assays/PK
SDP
Supplier:  Biochain Institute Inc Z5030017
Encompass_Preferred
BioChain's phosphate assay kit is a simple, direct and automation-ready assay for measuring phosphate concentration in biological and environmental samples without pretreatment. The improved Malachite Green method utilizes the malachite green dye and molybdate, which forms a stable colored complex specifically with inorganic phosphate. The intensity of the color is directly proportional to the phosphate concentration in the sample. The optimized formulation substantially reduces interference by substances in the raw samples.
Catalog No. 50-179-793
Explore available promotions