Learn More
Biochain Institute Inc Glucose Assay Kit, 200 Assays/PK
Supplier: Biochain Institute Inc Z5030025
BioChain's glucose concentration assay kit is a simple, direct and automation-ready assay for measuring glucose in serum or plasma without any pretreatment. The improved o-toluidine method utilizes a specific color reaction with glucose. The intensity of the color is directly proportional to the glucose concentration in the sample. The absorbance at 630nm is directly proportional to glucose concentration in the sample.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.