Learn More
Biochain Institute Inc Chloride Assay Kit, 500 assays/PK
Supplier: Biochain Institute Inc Z5030015
BioChain's chloride assay kit is a simple, direct and automation-ready assay for measuring chloride concentration in biological samples without pretreatment. The improved Fried method utilizes mercuric 2,4,6-tripyridyl-s-triazine, which forms a colored complex specifically with chloride. The intensity of the color is directly proportional to the chloride concentration in the sample. The optimized formulation substantially reduces interference by substances in the raw samples.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.