Learn More
Biochain Institute Inc Heme Assay Kit, 250 Assays/PK
SDP
Supplier:  Biochain Institute Inc Z5030027
Encompass_Preferred
BioChain's heme concentration assay kit is a simple, direct and automation-ready assay for measuring heme. The improved aqueous alkaline solution method, in which the heme is converted into a uniform colored form. The intensity of the color is directly proportional to the heme concentration in the sample. The optimized formulation exhibits high sensitivity and substantially reduces interference by substances in the raw samples.
Catalog No. 50-179-802
Explore available promotions