Learn More
Biochain Institute Inc Creatinine Assay Kit, 500 Assays/PK
Supplier: Biochain Institute Inc Z5030020
BioChain's creatinine concentration assay kit is a simple, direct and automation-ready assay for measuring creatinine in biological samples without pretreatment. The improved Jaffe method utilizes picrate that forms a red colored complex specifically with creatinine. The intensity of the color is directly proportional to the creatinine concentration in the sample. The optimized formulation substantially reduces interference by substances in the raw samples.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.