Learn More
Biochain Institute Inc Magnesium Assay Kit, 250 Assays/PK
SDP
Supplier:  Biochain Institute Inc Z5030032
Encompass_Preferred
BioChain's magnesium concentration assay kit is a simple, direct and automation-ready assay for measuring magnesium in biological samples without any pretreatment. A calmagite dye in the kit forms a colored complex specifically with magnesium. The intensity of the color is directly proportional to the magnesium concentration in the sample. The optimized formulation minimizes interference by potential substances.
Catalog No. 50-179-803
Explore available promotions