Learn More
Electron Microscopy Sciences Via-Cell Via-Cell Bioaerosol Sampling Cassette
SDP
Supplier:  Electron Microscopy Sciences 6521210
Encompass_Preferred
The Via-Cell disposable cassette for air sampling is designed to collect both viable and non-viable bioaerosols, maintain visibility of the viable mold spores and prevent growth during transport to the laboratory. It is allows analysis by culturing, PCR and direct microscopy.
To use simply connect it o any pump calibrated to 15 LBM and run your sample.
Catalog No. 50-338-49