Learn More
Chemglass Life Sciences 3L Unjacketed Bio Reactor Vessel
Supplier: Chemglass Life Sciences CLS138001
3L Unjacketed Bio Reactor Vessel. Vessels are constructed of heavy wall tubing and are graduated in 200mL subdivisions with numbers every 1000mL. Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors. Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.