Learn More
Bio Plas™ Uni-Flex™ Safety Caps
Protects laboratory staff from infectious aerosols
Supplier: Bio Plas™ 6575
Description
Protects laboratory staff from infectious aerosols
- Liquid-tight seal ensures sample integrity and prevent spillage of serum samples
- Designed to hold under centrifugation, agitation, refrigeration and freezing
- Stretchable, unique, over-locking design to prevent evaporation
- One-hand application and removal
- Trace-metal free material
Specifications
Liquid-tight seal | |
Polyethylene | |
12 mm culture or 13 mm blood collection tubes |
Safety Cap | |
Gray |