Learn More
Bio Plas™ Uni-Flex™ Safety Caps
Protects laboratory staff from infectious aerosols
Supplier: Bio Plas™ 6560
missing translation for 'nonReturnableMessage'
missing translation for 'returnPolicyLabel'
Description
Protects laboratory staff from infectious aerosols
- Liquid-tight seal ensures sample integrity and prevent spillage of serum samples
- Designed to hold under centrifugation, agitation, refrigeration and freezing
- Stretchable, unique, over-locking design to prevent evaporation
- One-hand application and removal
- Trace-metal free material
Specifications
Liquid-tight seal | |
Polyethylene | |
12 mm culture or 13 mm blood collection tubes |
Safety Cap | |
Lavender |
missing translation for 'provideContentCorrection'
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'
Spot an opportunity for improvement?
missing translation for 'provideContentCorrection'