Learn More
Research Products International Corp Biohazard Labmat, Safety Orange, 20 Inches Wide x 50 Feet Long per Roll
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Research Products International Corp 624675
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Labmat simplifies clean-up and protects surfaces from stains and spills. Safety Orange for biohazard designated use. Flexible polyethylene film backing prevents solutions from soaking through. The highly absorbent top surface traps water, dyes, corrosive chemicals and radioactive solutions. The entire sheet can be curled up and discarded after contamination. Rolls are easily cut with scissors while roll is retained in dispenser.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.