Learn More
Cole-Parmer Traceable Four-Channel Alarm Timer with Calibration
Supplier:  Cole-Parmer 9022535
Encompass_Preferred
This timer allows you to count up to or down from 100 hours with 1-second resolution. This timer features a 3/4" high display and multiple channels that can be used simultaneously. Each channel has a distinctive alarm that sounds for 1 minute, pressing any button will turn off the alarm. Timer memory recalls previous time set. Features time-of-day in 12- or 24-hour formats. Traceable Products have an individually-numbered Traceable Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
Catalog No. NC2063323
Please call customer service for current pricing. \n1-800-766-7000
Explore available promotions