Learn More
Electron Microscopy Sciences Beyer Bone Rongeur, Double Action
Supplier: Electron Microscopy Sciences 77987C
Double action bone rongeur with a curved jaw. The jaw width is 3.5 mm and are 7" long. Made from stainless steel with a polished finish