Learn More
Thomas Scientific Small Waste Container-Hinged Lid
SDP
Supplier:  Thomas Scientific 1226L57
Encompass
IBI’s BETA-GARD™ waste containers are designed for radioactive waste or any other laboratory waste that requires containment. These safe storage containers may be used with disposable NRC-approved waste bags. There are four (4) stationary waste containers available in convenient sizes with hinged lids, and one (1) large mobile waste container with heavy duty casters and a large handle for easy mobility in your lab. All BETA-GARD™ waste containers are constructed of heavy duty 1/2” cast acrylic for maximum protection of laboratory personnel.
Catalog No. 50-146-1148