Learn More
Thomas Scientific Medium Waste Container-Hinged Lid
Supplier: Thomas Scientific 1226L59
IBI’s BETA-GARD™ waste containers are designed for radioactive waste or any other laboratory waste that requires containment. These safe storage containers may be used with disposable NRC-approved waste bags. There are four (4) stationary waste containers available in convenient sizes with hinged lids, and one (1) large mobile waste container with heavy duty casters and a large handle for easy mobility in your lab. All BETA-GARD™ waste containers are constructed of heavy duty 1/2” cast acrylic for maximum protection of laboratory personnel.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.