Learn More
Research Products International Corp Beta RackBox with Two 4-Way Flipper Racks, 3/8 Inches Thick, Clear Acrylic
SDP
Supplier:  Research Products International Corp 145751
Encompass_Preferred
Beta RackBox provides convenient access to tubes when working with radioactive samples. Removable end panel allows rack to slide in or out. Unique hinged lid can be positioned at any angle to serve as a frontal shield. Two 4-Way Flipper racks are included. Constructed of heavy duty 3/8" thick clear acrylic for shielding 32P beta emissions. Safe for freezer storage to -70?C.
Catalog No. 50-213-042