Learn More
Sigma Aldrich Fine Chemicals Biosciences BETA-Nicotinamide mononucleotide, 1094-61-7, MFCD00038748, 25 mg
Supplier: Sigma Aldrich Fine Chemicals Biosciences N350125MG
Empirical Formula (Hill Notation): C11H15N2O8P, Molecular Weight: 334.22, ≥95% (HPLC), storage temp: −20°C, Synonym: β-NMN, β-Nicotinamide ribose monophosphate, NMN, Nicotinamide ribotide, Nicotinamide-1-ium-1-β-D-ribofuranoside 5′-phosphate.