Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHOWA™ 6784 Chemical Resistant Neoprene Gloves, Cotton Liner
Click to view available options
14 in. (36 cm) Long; Smooth
Description
- Chemical resistant against acids, caustics, oils, greases and many solvents
- Protection against physical hazards such as abrasion and cut
- Rough grip for wet applications
- Cotton liner keeps cool and comfortable in hot conditions or warm in cold conditions
- Ideal for biotechnology, cytostatic, life sciences, pharmaceuticals and API
Specifications
Certifications/Compliance | 21 CFR 170-199 |
Certifications/Compliance (Europe) | Cat III CE 0321, EN 407:4342XX |
Color | Black |
Latex Free | Yes |
Material | Neoprene |
Size | 10 |
EN388 Standard | EN 388:3111X |
EN 421 | No |
EN ISO 374-1 | Type A |
EN ISO 374-1 CHEMICAL LETTER CODES | AJKLMT |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction