Learn More
Berkshire™ EasyClean™ 360 Isolator Cleaning Tool
SDP
The perfect cover for a great cleaning tool
$30.45 - $1328.00
Specifications
For Use With (Application) Cleaning
View More Specs
Products 4
Catalog Number Mfr. No. Certifications/Compliance Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Certifications/Compliance Product Type Price Quantity & Availability  
17-987-360
SDP
View Documents
Berkshire™
EC360.ICT.1
ISO Class 3 to 8, EU Grade A/B, USP Isolator Cleaning Tool
Each for $430.50
Edge
Add to Cart
17-987-362
SDP
View Documents
Berkshire™
EC360.ECST.20
ISO Class 3 to 8, EU Grade A/B, USP Isolator Cleaning Tool Cover
Case of 220 for $1,328.00
Add to Cart
17-987-361
SDP
View Documents
Berkshire™
EC360.EC.6
ISO Class 3 to 8, EU Grade A/B, USP Isolator Cleaning Tool Cover
Pack of 26 for $181.70
Case of 6 PK for $962.00
Edge
Add to Cart
17-987-363
SDP
View Documents
Berkshire™
EC360.NW.40
ISO Class 5 to 8, EU Grade Isolator Cleaning Tool Cover
Pack of 5 for $30.45
Case of 40 PK for $1,035.00
Edge
Add to Cart
Description
Description
EasyClean™ TM 360 Elastic Isolator Cleaning Tool Covers Features Include:
  • Made from Berkshire's Super Polx TM 1200 continuous filament polyester wiping material
  • Laundered and packaged in an ISO Class 4 (Class 10) cleanroom for superior fiber, particle, and residue performance
  • Stretch fabric and elastic edge allow easy installation and secure fit on the mop head
  • Absorbent knit is abrasion resistant and durable to a wide range of cleaning solutions
  • Autoclavable
  • Non-Sterile and validated sterile versions available
EasyClean™ TM 360 Padded Isolator Cleaning Tool Covers Features Include:
  • 100% continuous filament knitted padded covers are laundered and packaged in an ISO Class 4 (Class 10) clean-room
  • Foam-free cushioned design ensures even surface coverage with reduced residues and contaminants
  • Easy-to-install covers fit securely and stay in place during use
  • Autoclavable
EasyClean™ 360 Nonwoven Isolator Cleaning Tool Covers features include:
  • Double-ply 55% polyester/45% cellulose
  • 100% nonwoven rayon inner layer for added absorbency
  • Elastic edge allows secure fit on the mop head during use
  • Autoclavable
Specifications
Specifications
Cleaning