Learn More
Cambridge Isotope Laboratories BENZOIC ACID CALIBRANT FOR QNMR 15mM BENZOIC ACID IN DMSO-D6 (120 UL FILL VOLUME), 1 3MM TUBE
Supplier: Cambridge Isotope Laboratories DLM7061E
BENZOIC ACID CALIBRANT FOR QNMR 15mM BENZOIC ACID IN DMSO-D6 (120 UL FILL VOLUME), 1 3MM TUBE