Learn More
Perkin Elmer US LLC Benzene In Gasoline Application Pack
Supplier: Perkin Elmer US LLC L1608012
The application pack contains: Two-port Leur Lock %T cell with KBr windows, 0.05 mm path length Spare 0.05 mm Teflon spacer for the %T cell, Qty. 6 2 mL Leur Lock syringes, Qty. 2 Liquid cell Leur connected waste tubing Fuels Touch Apps Software Kit AVI Manual Kit The Touch Apps Software Kit includes the software and SOPs to support the analysis of biofuels and provide a simplified workflow-driven operating environment; for faster, more reproducible analyses.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.