Learn More
Apexbio Technology LLC Benfotiamine,250mg CAS# 22457-89-2
Supplier: Apexbio Technology LLC A3236250
Benfotiamine, the lipid-soluble thiamine derivative used as a treatment for diabetic neuropathy, can inhibit three major pathways(the hexosamine pathway, the advanced glycation end product (AGE) formation pathway and the diacylglycerol (DAG)?protein kinase C (PKC) pathway)of hyperglycemic damage and prevent experimental diabetic retinopathy. Benfotiamine is a synthetic S-acyl derivative of thiamine (vitamin B1) for treating sciatica and other painful nerve conditions. More effective at increasing thiamin levels in blood and tissues than water-soluble salts like the previous vitamin B1. Other sizes are also available. Please inqury us for quote.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.