Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dynamic Diagnostics Benchtop Corner Shelf
Maximizes the available corner space of benches or tables
Supplier: Dynamic Diagnostics 4000147
Description
- Transparent design for easy visualization of everything stored underneath the shelf
- Assists with laboratory organization
Specifications
Corner Shelf | |
12 x 12 x 5.25 in. | |
Bench Organization | |
12 in. |
Not Autoclavable | |
Transparent | |
Medium Bench Height | |
Acrylic |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction