Learn More
World Precision Instrument Benchtop Autoclave, 16 Liter-ST5189-EU
Supplier: World Precision Instrument ST5189EU
Benchtop Autoclave ideal for small scale sterilization of microdissection and veterinary instruments.
The autoclave comes pre-programmed for basic sterilization. Choose either the 120ºC or 134ºC temperature mode, and the cycle time is fixed. This is the perfect autoclave for small spaces.
The stainless steel sterilization chamber accommodates a variety of liquids, media, instruments, glassware and plasticware. A mechanical and electrical safety interlock prevents the door from being opened until the pressure is released (0 PSI).
Simply press the Start button, and the entire sequence runs–fill, sterilize, exhaust and dry.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.