Learn More
OXFORD LAB PRODUCTS BenchMate CH24V Hematocrit Centrifuge
Supplier:  OXFORD LAB PRODUCTS CH24V
Encompass_Preferred
This affordable and compact hematocrit centrifuge is ideal for any laboratory determining erythrocyte fractions in blood and separating micro solutions. Safety is emphasized with the Oxford Lab Products BenchMate Hematocrit Centrifuge, equipped with an imbalance detection safety system, with an auto cut-off feature, and the lid lock safety feature. The lid of this hematocrit centrifuge releases automatically after run completion.
Catalog No. NC1686982
Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative.