Learn More
Laboratory Supply Network Inc Benchmark Scientific PlateFuge Microplate Centrifuge
Supplier: Laboratory Supply Network Inc BMS:C2000
Benchmark's PlateFuge, is the first "mini" sized centrifuge with a swing out rotor for microplates. Centrifugation of plates before PCR ensures all reactants are in the bottom of the wells for optimized concentrations and imporved yields. In spite of its remarkably small footprint (9 x 10in.), the PlateFuge includes a uniquely designed, swing out rotor (patent pending) capable of securing 2 microplates. The easily accessible rotor chamber includes two plate carriers that rest at a 75° angle. This allows plates to be insterted confidently without sealing tapes or caps. Upon closing the lid of the centrifuge, the rotor accelerates and the centrifugal force "swings" the plates into a horizontal position. Any droplets on thw walls of the plate, quickly concentrate into the well bottoms. With a g-force of over 400 xg, most samples can be spun down in less than 20 seconds.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.