Learn More
Bellco Glass, Inc. Heating blankets are designed to be used with a temperature controller and rtd temperature sensor for heating media inside a vessel to temperature. Male iec connector for use with temperature controllers Heating at 5 watts/sq.in. Flask size: 3/6L
Supplier: Bellco Glass, Inc. 196451040
Heating blankets are designed to be used with a temperature controller and rtd temperature sensor for heating media inside a vessel to temperature. Male iec connector for use with temperature controllers Heating at 5 watts/sq.in. Flask size: 3/6L