Learn More
Bellco Glass, Inc. Manufactured from type 1 class a Borosilicate glass. Bottom indent and 3 vertical baffles for optimal mixing. Sidearm cap: 45mm Center cap: 100mm Dimensions: 245" x 135" Flask size: 1l
SDP
Supplier:  Bellco Glass, Inc. 196581005
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Bottom indent and 3 vertical baffles for optimal mixing. Sidearm cap: 45mm Center cap: 100mm Dimensions: 245" x 135" Flask size: 1l
Catalog No. 50-194-5095
Explore available promotions