Learn More
Bel-Art™ SP Scienceware™ Portable Glove Box Accessory, Clamping Ring
Holds gloves in place. Bel-Art™ SP Scienceware™ Portable Glove Box Accessory, Clamping Rings are adjustable rings for use with sidENTRY (H50026-0000) glove boxes.
Supplier: Bel-Art™ H500290200/EMD
Description
- Fastens gloves firmly in glove ports
- Adjustable rings sold in sets of 2
- Fits any glove box with 6 in. diameter glove port holes, including Techni-Dome, Portable Glove Box or Clear View
Specifications
Portable Glove Box | |
Clamping Ring | |
Fasten Glove Firmly to Glove Box |