Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bel-Art™ SP Scienceware™ Accessories for Write-on Tape Dispenser
The U-shaped stainless steel adapter presents the dispenser at a 45° angle off the wall to facilitate writing
Supplier: Bel-Art™ F134730001
Description
- Economy Model Dispenser (Mfr. No. F13473-0000) can be wall mounted vertically or horizontally with the platform positioned to the left or right
- Compatible with Wet Surface Pen (sold separately), which offers consistent ink flow on vertical surfaces
Specifications
Wall Mount Foot | |
Stainless Steel | |
Mounts vertically or horizontally |
Rolls of Tape | |
4 Rolls x 25 mm W (1 in.) or 5 Rolls x 12 to 19 mm W (1/2 to 3/4 in.) |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title