Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher Science Education™ Beef Extract
$45.10
Specifications
Additive | Beef Extract |
---|---|
Form | Powder |
For Use With (Application) | Dehydrated culture media |
Catalog Number | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Quantity | Price | Quantity & Availability | ||||||
S25661A
|
100 g |
Each for $45.10
Case of 8 EA for $328.00
|
|
||||||
Specifications
Beef Extract | |
Powder | |
Dehydrated culture media |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title