Learn More
HemoCue America Beckman Coulter™ Gastroccult™ Gastric Sample Occult Blood Test Kit
Two tests in one
Supplier: HemoCue America 66140A
Description
- CLIA Complexity: Waived
- CPT Codes: 87723 for occult blood, 83986 for pH
- Slide test for detecting occult blood and determining pH in gastric samples, both aspirate and vomitus, on the same test card
- On-slide Performance Monitor assures that the occult blood test is performing to product specifications
- Scale of pH test ranges from a clinically useful 1.3 to 7.8
- Both test results are available within one minute
- Test cards, developer, and applicator straws sold separately
Specifications
Waived | |
Straw Applicator | |
40 Pack |