Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HemoCue America Beckman Coulter FlexSure™ HP Serum Test Kit
Simple three-step procedure
Supplier: HemoCue America 81050
Description
- CPT Code: 86677
- CLIA Complexity: Non-waived
- Test for IgG antibodies specific to H. pylori in serum
- Reverse-flow immunochromatography
- Test results in four minutes
- Visually read results
- Requires no specialized equipment or instrumentation
- Two visible control lines on the test strip indicate a positive test
- Compared to biopsies, relative sensitivity is 97% and overall agreement is 93%
- Compared to EIA tests, relative sensitivity is 95%, relative specificity is 94% and overall agreement is 95%
Order Info
Control kit ordered separately
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction