Learn More
ILEX LIFE SCIENCES LLC denovoMATRIX beadMATRIX - chemically-defined precoated microcarriers for MSCs culture, 10 g
SDP
Supplier:  ILEX LIFE SCIENCES LLC M0101
Encompass_Preferred
DESCRIPTION: Generate clinically relevant quantities of mesenchymal stromal cells (MSCs) with beadMATRIX. beadMATRIX are animal component-free, pre-coated microcarriers specially designed for bioreactor-scale expansion of MSCs in serum-free and xeno-free media. Manufactured in Germany by denovoMATRIX. HIGH YIELD, HIGH QUALITY: Application of beadMATRIX accelerates the proliferation of highly viable and therapeutically active MSCs up to 25% compared to the leading brand competitor. Harvested cells show MSC-specific characteristics as defined by the International Society of Cellular Therapy such as distinctive CD-marker expression and trilineage differentiation potential. In addition, they present immunomodulatory potential and angiogenic activity. SCALE-UP COMPATIBLE: beadMATRIX has been successfully tested with different bioreactor setups and several cell culture media.
Catalog No. NC2209155