Learn More
Electron Microscopy Sciences Beryllium Substrate Planchet, 50.8 mm x 1 mm
Supplier: Electron Microscopy Sciences 76016
These planchets are prepared from high purity beryllium (min. 98.5%) by electro-fusion to provide vacuum tight (~1x10-9atm-cm3/sec) and +/-10% dimensional tolerances.
Size: 50.8 mm x 1 mm