Learn More
BD Vacutainer™ Plastic Blood Collection Tubes No Additives: Conventional Stopper
Sterile plastic tubes with no additives for venous blood and urine collection
Supplier: BD 366704
Description
- Blood is drawn directly from the vein into the evacuated sterile collection tubes
- Only one venipuncture is needed to draw multiple samples
- Tubes can be changed while needle and plastic holder remain in place on patient's arm
- Closures are red for easy identification
- Made of polyethylene terephthalate (PET)
- Provide a safer work environment for all personnel by (1) reducing risk of tube breakage during handling; (2) eliminating breakage when tubes are centrifuged at higher speeds; and (3) eliminating breakage or chipping of tube rims when stopper is removed and reinserted
- In addition to reducing potential injuries, fewer tube breakages result in less workflow interruption and fewer patient redraws, providing smoother day-to-day operation
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
No Additive | |
Conventional Stopper | |
13 x 75 mm |
Plastic | |
Red/Light Gray | |
Paper |