Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD SurePrep™ Capillary Tubes
Specimen collection has never been easier or safer
$167.00
Specifications
Sterility | Non-sterile |
---|---|
Closure Type | Self-Sealing Plug |
For Use With (Application) | Capillary Blood Collection Tube |
Description | Size: 1.1 X 75 mm |
Catalog Number | Mfr. No. | Additive | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Additive | Quantity | Price | Quantity & Availability | ||||
14-170-9B
|
BD
420316 |
Heparin | 2 Vials of 100 Tubes Each |
Pack of 200 for $167.00
|
![]() |
14-170-9C
|
BD
420317 |
Heparin | 10 Vials of 100 Tubes Each |
Pack of 1000 for $833.00
|
N/A |
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
14-170-9
|
BD
420314 |
Plain | 2 Vials of 100 Tubes Each |
Pack of 200 for $167.00
|
![]() |
14-170-9D
|
BD
420318 |
Heparin (Precalibrated) | 2 Vials of 100 Tubes Each |
Pack of 200 for $167.00
|
|
22-307781
|
BD
420315 |
Plain | 10 Vials of 100 Tubes Each |
Pack of 1000 for $833.00
|
N/A |
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
- Tubes feature self-sealing plug and multilayered Mylar™ wrap
- Reduce exposure to HIV, HBV, and other bloodborne pathogens
- A channel in the system allows air to escape during filling; once the sample touches the plug, the channel seals automatically
- Simply fill the tube, hold vertically, then centrifuge and read
- In the event of accidental breakage, the protective Mylar wrap helps contain blood and minimizes exposure to glass fragments
Specifications
Non-sterile | |
Self-Sealing Plug | |
Capillary Blood Collection Tube | |
Size: 1.1 X 75 mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title