Learn More
BD Human Th1/Th2/Th17 Phenotyping Kit
Simultaneously view Th1, Th2 and Th17 cells in mouse model systems
Supplier:  BD 560751
Catalog No. BDB560751
Add to Cart
Includes: Human Th1/Th2/Th17 Phenotyping Cocktail (CD4 PerCP-Cy™ 5.5, IL-17A PE, IFN-γ FITC, IL-4 APC), BD Cytofix™ Fixation Buffer, BD Perm/Wash™ Buffer, BD GolgiStop™ Protein Transport Inhibitor
Description
Description
  • Provide a simple solution for measurement of multiple cytokines or transcriptions factors from a single sample using multicolor flow cytometry
  • Can be used to analyze ex vivo lymphoid cell samplesor to monitor T helper cell differentiation by cells cultured within various experimental model systems
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1 Kit
Product Suggestions
Product Suggestions
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For research use only. Not for diagnostic or therapeutic procedures.