Learn More
BD Directigen™ S. pneumoniae Test Kit
Supplier: BD 251960
This item is not returnable.
View return policy
Description
Presumptive latex slide agglutination test for the qualitative detection of antigens to S. pneumoniae directly in cerebrospinal fluid (CSF), serum or urine.Test kit provides confirmation and serogrouping capabilities from suspected colonies of S. pneumoniae.
Visible agglutination occurs when a sample containing these bacterial antigens is reacted with its respective antibody-coated latex beads.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
High, Moderate | |
S. pneumoniae Kit | |
90 Tests |
Blood Culture, CSF, Serum, Urine | |
One bottle S. pneumoniae Antibody 1mL, R1, one bottle positive control 1.5mL, one bottle negative control 1.5mL, one bottle control Latex reagent A stirrers 0.5mL, Plastic | |
15 to 20 min. |