Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD Difco™ Lactose Monohydrate
Media Additive; Microbiology; Lactose Monohydrate; Quantity: 500g, Difco
Supplier: BD 215620
Description
Media Additive; Microbiology; Lactose Monohydrate; Quantity: 500g, Difco
Specifications
ISO/DIS, 11014 | |
Solid | |
Bottle |
Lactose Monohydrate | |
Dehydrated culture media | |
500 g |
Safety and Handling
ShelfLife : 1825 Days
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title