Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BD™ Difco™ Dextrose Glucose, Anhydrous
Used with microbiological culture media. BD™ Difco™ Dextrose Glucose, Anhydrous is a form of D(+)-Glucose. The dehydrated form provides greater convenience for the laboratorian and bioprocessing specialist.
Supplier: BD 215510
Description
D-Glucose is a simple sugar that circulates in the blood of animals as blood sugar, and is made in plants during photosynthesis. It is an important source of power for cellular respiration. Glucose is stored as as a starch in plants and as glycogen in animals.
- CAS: 50-99-7
- EC No.: 200-075-1
- Chemical formula: C6H12O6
- Molar mass: 180.16g/mol
- Synonym: Blood Sugar
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
ISO/DIS, 11014 | |
<30°C | |
Anhydrous Solid | |
Pail |
Whitish | |
Dextrose Glucose | |
Microbiological culture media | |
2 kg |
Safety and Handling
ShelfLife : 1460 Days
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction