Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD Difco™ Buffered Listeria Enrichment Broth Base
Enrichment broth for cultivation of Listeria spp. from food
Supplier: BD 214919
Specifications
ISO/DIS, 11014 | |
<30°C | |
United States | |
Pancreatic digest of Casein 17g/L, Soytone 3g/L, Dextrose 2.5g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 2.5g/L, Disodium Phosphate 9.6g/L, Monopotassium Phosphate 1.35g/L, Yeast extract 6g/L, Sodium Pyruvate 1.1g/L | |
Pail |
Light Beige | |
Solid | |
Buffered Listeria enrichment broth base | |
For enrichment of Listeria | |
2000 g |
Safety and Handling
ShelfLife : 1460 Days
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title