Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD D- Luciferin Sodium Salt
Specific for firefly luciferase in the illustrated reactions
$320.00 - $562.00
Specifications
Formulation | Lyophilized |
---|---|
Kit Contents | Sodium salt |
Type | D-Luciferin |
Content And Storage | -20°C |
For Use With (Application) | Luciferase assay |
Includes: Sodium salt
Description
Purified by HPLC techniques to remove dehydroluciferin and L-Luciferin, both of which are competitive inhibitors of firefly luciferase. Form dehydroluciferin in presence of ultraviolet light and/or moisture
- Synthetically made substrate
- lyophilized, in monosodium salt form
- Non-buffered solution
- Reconstitute lyophilized D-Luciferin with deionized water
Specifications
Lyophilized | |
D-Luciferin | |
Luciferase assay |
Sodium salt | |
-20°C |
Research use only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title