Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Prepared Plated Media: Trypticase™ Soy Agar (Soybean-Casein Digest Agar)
Used for isolation and cultivation of non-fastidious and fastidious microorganisms
Supplier: BD 221185
This item is not returnable.
View return policy
Description
- Ready for immediate use
- Not used for anaerobes
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
ISO/DIS, 11014 | |
2°C to 8°C | |
Trypticase™ Soy Agar (Soybean Casein digest Agar) | |
Cultivation of Non-fastidious and fastidious microorganisms (not for anaerobes) | |
20/Pk. |
Light Beige | |
United States | |
Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Sodium Chloride 5g/L, Agar 15g/L | |
Plate | |
100 x 15 mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction