Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Prepared Plated Media: Mueller Hinton II Agar
For antimicrobial disc diffusion susceptibility testing of common, rapidly growing bacteria by the Bauer-Kirby method
Supplier: BD 221994
This item is not returnable.
View return policy
Description
- Ready-to-use
- 100 x 10mm, 100 x 15mm and 150 x 15mm-style plates
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
ISO/DIS, 11014 | |
2°C to 8°C | |
United States | |
Beef extract 2g/L, Acid Hydrolysate of Casein 17.5g/L, Starch 1.5g/L, Agar 17g/L | |
150 x 15mm Plate |
Beige | |
Solid | |
Mueller Hinton II Agar | |
Antimicrobial disc diffusion susceptibility testing of common rapidly growing bacteria by the Bauer-Kirby method | |
8/Pk. |
Safety and Handling
ShelfLife : 98 Days
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction