Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Mycoflask™ Prepared Lim Broth (Todd Hewitt with CNA) Media
For selective enrichment of group B Streptococci (Streptococcus agalactiae), especially from genital specimens
$317.00
Specifications
Content And Storage | 2°C to 8°C |
---|---|
Form | Liquid |
Manufacturing Origin | United States |
Media Type | Lim Broth |
Formulation | Heart infusion from (solids) 3.1g/L, Peptone 20g/L, Dextrose 2g/L, Sodium Chloride 2g/L, Disodium Phosphate 0.4g/L, Sodium Carbonate 2.5g/L, Yeast extract 10g/L, Colistin 0.01g/L, Nalidixic Acid 0.015g/L |
Catalog Number | Mfr. No. | Certifications/Compliance | Color | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Certifications/Compliance | Color | Quantity | Price | Quantity & Availability | |||
L96266
|
BD
296266 |
CE, BS12, CMPH2, MCM9 | Beige | 5 mL |
Pack of 100 for $317.00
|
|
|||
L4392209
|
BD
292209 |
ISO/DIS, 11014, BS12, CMPH2, MCM9 | Yellow | 10/Pk. | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
Description
- Available as K tubes
Specifications
2°C to 8°C | |
United States | |
Heart infusion from (solids) 3.1g/L, Peptone 20g/L, Dextrose 2g/L, Sodium Chloride 2g/L, Disodium Phosphate 0.4g/L, Sodium Carbonate 2.5g/L, Yeast extract 10g/L, Colistin 0.01g/L, Nalidixic Acid 0.015g/L | |
Tube |
Liquid | |
Lim Broth | |
For selective enrichment of group B Streptococci, especially from genital specimens |
Safety and Handling
ShelfLife : 274 Days
For in vitro diagnostic use
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title