Learn More
BD BBL™ Enterotube™ II Tube Test
Supplier: BD 211832
This item is not returnable.
View return policy
Description
- Prepared sterile multimedia tube for rapid differential identification of Gram-negative bacteria (Enterobacteriaceae)
- Self-contained, sterile, compartmented plastic tube contains twelve different conventional media plus a self-contained inoculating wire
- Subsequent performance of 15 biochemical tests (glucose, gas production, lysine decarboxylase, ornithine decarboxylase, H2S, indole, adonitol, lactose, arabinose, sorbitol, Voges-Proskauer, dulcitol, phenylalanine deaminase, urea and citrate) from a single bacterial colony together with the Interpretation Guide (order separately) allow identification of Enterobacteriaceae
- Innoculation of all 12 media occurs simultaneously
Order Info
Package contains 25 tubes.