Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Mycoflask™ Trypticase™ Soy Broth Prepared Media
Used for the cultivation of fastidious and nonfastidious microorganisms from a variety of clinical and nonclinical specimens
Supplier: BD 221715
Description
Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.
Specifications
CE, AOAC, BAM, BS12, CCAM, CLSI, CMPH2, COMPF, EP, EPA, ISO, JP, MCM9, SMD, USDA, USP | |
15°C to 25°C | |
United States | |
Pancreatic digest of Casein 17g/L, Papaic digest of Soybean Meal 3g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 2.5g/L, Dextrose 2.5g/L | |
Size K tubes with cap |
Beige | |
Liquid | |
Trypticase Soy Broth | |
For qualitative procedures for the cultivation of fastidious and non-fastidious microorganisms | |
10 x 5 mL |
Safety and Handling
ShelfLife : 548 Days
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For in vitro diagnostic use.
Spot an opportunity for improvement?
Provide Content Correction