Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bay 11-7085, Tocris Bioscience™

Catalog No. 174310
Click to view available options
Quantity:
10 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Irreversible inhibitor of TNF-α-induced IκBα phosphorylation

Irreversible inhibitor of TNF-α-stimulated IκBα phosphorylation (IC50 ∽ 10 μM); leads to decreased NF-κB and subsequent decreased expression of adhesion molecules. Also reversibly activates MAP kinases and stimulates apoptosis. Anti-inflammatory in vivo.

Chemical Identifiers

CAS 196309-76-9
Molecular Formula C13H15NO2S
Molecular Weight (g/mol) 249.328
InChI Key VHKZGNPOHPFPER-ONNFQVAWSA-N
Synonym e-3-4-tert-butylphenylsulfonyl acrylonitrile, e-3-4-tert-butyl phenyl sulfonyl acrylonitrile, e-3-4-t-butylphenylsulfonyl-2-propenenitrile, e-3-4-tert-butylphenyl sulfonylprop-2-enenitrile, e 3-4-t-butylphenyl sulfonyl-2-propenenitrile, 2-propenenitrile, 3-4-1,1-dimethylethyl phenyl sulfonyl-, 2e, 2e-3-4-tert-butylbenzenesulfonyl prop-2-enenitrile, bay hplc , solid, e-3-4-tert-butyl-benzenesulfonyl-acrylonitrile, e-3-4-tert-butylphenyl sulfonyl-2-propenenitrile
PubChem CID 5353432
IUPAC Name (E)-3-(4-tert-butylphenyl)sulfonylprop-2-enenitrile
SMILES CC(C)(C)C1=CC=C(C=C1)S(=O)(=O)C=CC#N

Specifications

Quantity 10 mg
Formula Weight 249.33
Percent Purity >99%
Chemical Name or Material Bay 11-7085
Recommended Storage Store at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.